4-(Trifluoromethylsulphonyl)benzonitrile
Supplier: Apollo Scientific
Warning
PC0520-5G PC0520-1G
APOSPC0520-5GEA 217 EUR
APOSPC0520-5G APOSPC0520-1G
4-(Trifluoromethylsulphonyl)benzonitrile
4-(Trifluoromethylsulphonyl)benzonitrile
Formula: C₈H₄F₃NO₂S
MW: 235.18 g/mol
Melting Pt: 87…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01631631
CAS Number: 312-21-0
UN: 3439
ADR: 6.1 III