4-Chloro-2-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Warning
PC0558-25G PC0558-5G PC0558-100G
APOSPC0558-25GEA 84.1 EUR
APOSPC0558-25G APOSPC0558-5G APOSPC0558-100G
4-Chloro-2-(trifluoromethyl)benzaldehyde
4-Chloro-2-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
MW: 208.57
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD03092987
CAS Number: 320-43-4