2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole-4-carbaldehyde
PC0563-1G PC0563-5G PC0563-10G
APOSPC0563-1GEA 24.8 EUR
APOSPC0563-1G APOSPC0563-5G APOSPC0563-10G
2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde 97%
2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde
Formula: C₈H₄F₂O₃
MW: 186.11 g/mol
Boiling Pt: 62…64 °C
Flash Pt: 91.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00792419
CAS Number: 119895-68-0