(S)-(-)-4-Fluoro-α-methylbenzylamine
Supplier: Apollo Scientific
Danger
Synonyms: (S)-1-(4-Fluorophenyl)ethanamine, (S)-(-)-1-(4-Fluorophenyl)ethylamine
PC0613-25G PC0613-5G
APOSPC0613-25GEA 603 EUR
APOSPC0613-25G APOSPC0613-5G
(S)-(-)-4-Fluoro-α-methylbenzylamine
(S)-(-)-4-Fluoro-α-methylbenzylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 76 °C/22mmHg
Melting Pt: –30 °C
Density: 1.03 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03092999
CAS Number: 66399-30-2
UN: 2735
ADR: 8 II