(S)-(+)-1,1'-Binaphthyl-2,2'-diylbis(trifluoromethanesulphonate)
Supplier: Apollo Scientific
Danger
Synonyms: (S)-(+)-1,1'-Bi-2-naphthol bis(trifluoromethanesulphonate), (S)-[1,1-Binaphthalene]-2,2-diyl bis(trifluoromethanesulphonate), (S)-(+)-1,1'-Bi-2-naphthol Bis(trifluoromethanesulphonate)
PC0615-25G
APOSPC0615-25GEA 195 EUR
APOSPC0615-25G
(S)-(+)-1,1'-Binaphthyl-2,2'-diylbis(trifluoromethanesulphonate)
(S)-(+)-1,1'-Binaphthyl-2,2'-diylbis(trifluoromethanesulphonate)
Formula: C₂₂H₁₂F₆O₆S₂
MW: 550.44 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00274615
CAS Number: 128544-05-8
UN: 3261
ADR: 8,II