1-Bromo-2-fluoro-4-iodobenzene 98%
Supplier: Apollo Scientific
Warning
PC0670-500G PC0670-100G PC0670-1KG
APOSPC0670-500GEA 221 EUR
APOSPC0670-500G APOSPC0670-100G APOSPC0670-1KG
1-Bromo-2-fluoro-4-iodobenzene 98%
1-Bromo-2-fluoro-4-iodobenzene
Formula: C₆H₃BrFI
MW: 300.9 g/mol
Boiling Pt: 80 °C/2.5mmHg
Melting Pt: 34…38 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042577
CAS Number: 136434-77-0