4-Fluorocatechol 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Fluoro-1,2-dihydroxybenzene, 4-Fluoropyrocatechol
PC0672-1G PC0672-5G PC0672-500MG
APOSPC0672-1GEA 125 EUR
APOSPC0672-1G APOSPC0672-5G APOSPC0672-500MG
4-Fluorocatechol 97%
4-Fluorocatechol
Formula: C₆H₅FO₂
MW: 128.10 g/mol
Melting Pt: 90…91 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01317539
CAS Number: 367-32-8
EINECS: 226-356-9
UN: 1759
ADR: 8,II