(Difluoromethyl)benzene 95%
Supplier: Apollo Scientific
Danger
PC0683-1G PC0683-25G PC0683-5G
APOSPC0683-1GEA 40.6 EUR
APOSPC0683-1G APOSPC0683-25G APOSPC0683-5G
(Difluoromethyl)benzene 95%
(Difluoromethyl)benzene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 133…139 °C
Density: 1.138 g/cm³ (25 °C)
Flash Pt: 35.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03788488
CAS Number: 455-31-2
UN: 2920
ADR: 8+3 II