N-(4-Fluorobenzylidene)aniline
Supplier: Apollo Scientific
Warning
PC0719-5G
APOSPC0719-5GEA 168 EUR
APOSPC0719-5G
N-(4-Fluorobenzylidene)aniline
N-(4-Fluorobenzylidene)aniline
Formula: C₁₃H₁₀FN
MW: 199.23 g/mol
Boiling Pt: 176 °C
Melting Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00017971
CAS Number: 5676-81-3