4-Bromo-2,6-difluorobenzoyl chloride
Supplier: Apollo Scientific
Danger
PC0738-1G PC0738-5G
APOSPC0738-1GEA 132 EUR
APOSPC0738-1G APOSPC0738-5G
4-Bromo-2,6-difluorobenzoyl chloride
4-Bromo-2,6-difluorobenzoyl chloride
Formula: C₇H₂BrClF₂O
Boiling Pt: 222…223 °C
Storage Temperature: Ambient
MDL Number: MFCD03094086
CAS Number: 497181-19-8
UN: 3265
ADR: 8,II