2,3,4,5-Tetrafluorobenzonitrile
Supplier: Apollo Scientific
Warning
PC0743-100G PC0743-25G PC0743-5G
APOSPC0743-100GEA 330 EUR
APOSPC0743-100G APOSPC0743-25G APOSPC0743-5G
2,3,4,5-Tetrafluorobenzonitrile
2,3,4,5-Tetrafluorobenzonitrile
Formula: C₇HF₄N
MW: 175.09 g/mol
Boiling Pt: 164…165 °C
Density: 1.463 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00040187
CAS Number: 16582-93-7
UN: 3276
ADR: 6.1,III