3,5-Dichloro-α,α,α-trifluorotoluene 97%
Supplier: Apollo Scientific
Danger
PC0797-25G
APOSPC0797-25GEA 137 EUR
APOSPC0797-25G
3,5-Dichloro-α,α,α-trifluorotoluene 97%
3,5-Dichloro-α,α,α-trifluorotoluene
Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
Boiling Pt: 173…174 °C
Melting Pt: –13…–12 °C
Density: 1.47 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00203929
CAS Number: 54773-20-5
UN: 1760
ADR: 8,III