3-Chloro-2,4-difluoroaniline 99%
Supplier: Apollo Scientific
|
Warning
|
PC0802-25G
PC0802-5G
APOSPC0802-25GEA
119
EUR
APOSPC0802-25G
APOSPC0802-5G
3-Chloro-2,4-difluoroaniline 99%
3-Chloro-2,4-difluoroaniline
Formula:
C6H4ClF2N MW: 163.5525 g/mol Melting Pt: 60…62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042200 CAS Number: 2613-34-5 |