2-Chloro-4-fluorophenylhydrazine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: (2-Chloro-4-fluorophenyl)hydrazine hydrochloride
PC0816-25G PC0816-5G PC0816-100G
APOSPC0816-25GEA 42.2 EUR
APOSPC0816-25G APOSPC0816-5G APOSPC0816-100G
2-Chloro-4-fluorophenylhydrazine hydrochloride
2-Chloro-4-fluorophenylhydrazine hydrochloride
Formula: C₆H₇Cl₂FN₂
MW: 197.0376 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788524
CAS Number: 497959-29-2