3-Fluoro-2-methoxybenzoic acid
Supplier: Apollo Scientific
Warning
PC0828-1G PC0828-5G PC0828-25G
APOSPC0828-1GEA 34.8 EUR
APOSPC0828-1G APOSPC0828-5G APOSPC0828-25G
3-Fluoro-2-methoxybenzoic acid
3-Fluoro-2-methoxybenzoic acid
Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 112…114 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00973972
CAS Number: 106428-05-1