Ethyl-5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
PC0837-10G PC0837-1G
APOSPC0837-10GEA 411 EUR
APOSPC0837-10G APOSPC0837-1G
Ethyl-5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Formula: C₁₂H₁₂FN₃O₂
MW: 249.2410 g/mol
Melting Pt: 153…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00173917
CAS Number: 138907-68-3