2,3-Difluoro-4-hydroxybenzoic acid
Supplier: Apollo Scientific
Warning
PC0867-1G PC0867-5G
APOSPC0867-1GEA 60.9 EUR
APOSPC0867-1G APOSPC0867-5G
2,3-Difluoro-4-hydroxybenzoic acid
2,3-Difluoro-4-hydroxybenzoic acid
Formula: C₇H₄F₂O₃
MW: 174.1 g/mol
Melting Pt: 205…211 °C
Storage Temperature: Ambient
MDL Number: MFCD01321116
CAS Number: 175968-39-5