2-Bromo-3-fluoroanisole 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-1-fluoro-3-methoxybenzene
PC0875-5G PC0875-100G PC0875-25G
APOSPC0875-5GEA 21.9 EUR
APOSPC0875-5G APOSPC0875-100G APOSPC0875-25G
2-Bromo-3-fluoroanisole 99%
2-Bromo-3-fluoroanisole
Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 220 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD03788540
CAS Number: 446-59-3