4-Chloro-2-(trifluoromethyl)phenylisothiocyanate 98%
Supplier: Apollo Scientific
Warning
PC0876-25G PC0876-5G
APOSPC0876-25GEA 531 EUR
APOSPC0876-25G APOSPC0876-5G
4-Chloro-2-(trifluoromethyl)phenylisothiocyanate 98%
4-Chloro-2-(trifluoromethyl)phenylisothiocyanate
Formula: C₈H₃ClF₃NS
MW: 237.63 g/mol
Boiling Pt: 118 °C/8mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00270100
CAS Number: 99195-86-5
UN: 3334
ADR: 9,III