3-Chloro-5-fluoroanisole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-3-fluoro-5-methoxybenzene
PC0904-10G PC0904-5G
APOSPC0904-10GEA 97.3 EUR
APOSPC0904-10G APOSPC0904-5G
3-Chloro-5-fluoroanisole 98%
3-Chloro-5-fluoroanisole
Formula: C₇H₆ClFO
MW: 160.58 g/mol
Boiling Pt: 189 °C
Density: 1.265 g/cm³ (25 °C)
Flash Pt: 79.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00070770
CAS Number: 202925-08-4