4'-Methoxy-2'-(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Methoxy-2-(trifluoromethyl)phenyl)ethanone
PC0913-1G PC0913-5G PC0913-10G PC0913-25G
APOSPC0913-1GEA 33.5 EUR
APOSPC0913-1G APOSPC0913-5G APOSPC0913-10G APOSPC0913-25G
4'-Methoxy-2'-(trifluoromethyl)acetophenone 98%
4'-Methoxy-2'-(trifluoromethyl)acetophenone
Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01091009
CAS Number: 220141-75-3