2-Fluoro-5-(hydroxymethyl)benzoic acid 97%
Supplier: Apollo Scientific
Warning
PC0943-1G PC0943-250MG
APOSPC0943-1GEA 346 EUR
APOSPC0943-1G APOSPC0943-250MG
2-Fluoro-5-(hydroxymethyl)benzoic acid 97%
2-Fluoro-5-(hydroxymethyl)benzoic acid
Formula: C₈H₇FO₃
MW: 170.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862001
CAS Number: 481075-38-1