3-Bromo-4-fluorophenol
Supplier: Apollo Scientific
Warning
PC0957-1G PC0957-25G PC0957-5G PC0957-100G
APOSPC0957-1GEA 14.7 EUR
APOSPC0957-1G APOSPC0957-25G APOSPC0957-5G APOSPC0957-100G
3-Bromo-4-fluorophenol
3-Bromo-4-fluorophenol
Formula: C₆H₄BrFO
Boiling Pt: 78 °C/0.5mmHg
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD03425884
CAS Number: 27407-11-0