2,2,2-Trifluoroethyl formate 95%
Supplier: Apollo Scientific
Danger
PC0989-25G PC0989-5G
APOSPC0989-25GEA 63.8 EUR
APOSPC0989-25G APOSPC0989-5G
2,2,2-Trifluoroethyl formate 95%
2,2,2-Trifluoroethyl formate
Formula: C₃H₃F₃O₂
Boiling Pt: 60 °C
Density: 1.317 g/cm³ (25 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD06797733
CAS Number: 32042-38-9
UN: 3272
ADR: 3,II