4-Amino-3-fluorobenzonitrile 98%
Supplier: Apollo Scientific
Danger
PC0999-25G PC0999-5G
APOSPC0999-25GEA 27.7 EUR
APOSPC0999-25G APOSPC0999-5G
4-Amino-3-fluorobenzonitrile 98%
4-Amino-3-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Melting Pt: 87…90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00055559
CAS Number: 63069-50-1
UN: 3439
ADR: 6.1 III