2,4-Difluoro-5-nitrobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC1001-25G PC1001-5G
APOSPC1001-25GEA 34.8 EUR
APOSPC1001-25G APOSPC1001-5G
2,4-Difluoro-5-nitrobenzonitrile 98%
2,4-Difluoro-5-nitrobenzonitrile
Formula: C₇H₂F₂N₂O₂
MW: 184.1000 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07772030
CAS Number: 67152-20-9
UN: 2811
ADR: 6.1 III