1,2,4,5-Tetrafluoro-3,6-(trifluoromethyl)benzene
Supplier: Apollo Scientific
Warning
Synonyms: Perfluoro(1,4-dimethylbenzene)
PC10025-1G PC10025-5G PC10025-25G
APOSPC10025-1GEA 65.4 EUR
APOSPC10025-1G APOSPC10025-5G APOSPC10025-25G
1,2,4,5-Tetrafluoro-3,6-(trifluoromethyl)benzene
1,2,4,5-Tetrafluoro-3,6-(trifluoromethyl)benzene
Formula: C₈F₁₀
MW: 286.07 g/mol
Boiling Pt: 121…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00054264
CAS Number: 651-89-8