Allylpentafluorobenzene 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(Pentafluorophenyl)-1-propene, 1-Allyl-2,3,4,5,6-pentafluorobenzene
PC1006-25G
PC1006-5G
PC1006-1G
APOSPC1006-25GEA
139
EUR
APOSPC1006-25G
APOSPC1006-5G
APOSPC1006-1G
Allylpentafluorobenzene 96%
Allylpentafluorobenzene
Formula:
(C₆F₅)CH₂CHCH₂ MW: 208.13 g/mol Boiling Pt: 148…149 °C Melting Pt: –64 °C Density: 1.358 g/cm³ (25 °C) Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000301 CAS Number: 1736-60-3 EINECS: 217-083-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...