Triethoxy(pentafluorophenyl)silane 95%
Supplier: Apollo Scientific
Warning
Synonyms: (Triethoxysilyl)pentafluorobenzene, Triethoxy(perfluorophenyl)silane
PC10076-5G PC10076-1G
APOSPC10076-5GEA 215 EUR
APOSPC10076-5G APOSPC10076-1G
Triethoxy(pentafluorophenyl)silane 95%
Triethoxy(pentafluorophenyl)silane
Formula: C₁₂H₁₅F₅O₃Si
MW: 330.33 g/mol
Boiling Pt: 68…69 °C/1.5mmHg
Density: 1.242 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD03411257
CAS Number: 20083-34-5