3-(But-1-yl)-1,2-dimethyl-1H-imidazol-3-ium hexafluorophosphate
Supplier: Apollo Scientific
Warning
PC10104-5G PC10104-25G
APOSPC10104-5GEA 21.9 EUR
APOSPC10104-5G APOSPC10104-25G
3-(But-1-yl)-1,2-dimethyl-1H-imidazol-3-ium hexafluorophosphate
3-(But-1-yl)-1,2-dimethyl-1H-imidazol-3-ium hexafluorophosphate
Formula: C₉H₁₇F₆N₂P
Melting Pt: 38…40 °C
Density: 1.2416 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04039211
CAS Number: 227617-70-1