2-Bromo-6-nitro-4-(trifluoromethoxy)aniline
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4-(trifluoromethoxy)-6-nitroaniline
PC10108-25G PC10108-5G
APOSPC10108-25GEA 199 EUR
APOSPC10108-25G APOSPC10108-5G
2-Bromo-6-nitro-4-(trifluoromethoxy)aniline
2-Bromo-6-nitro-4-(trifluoromethoxy)aniline
Formula: C₇H₄BrF₃N₂O₃
MW: 301.0200 g/mol
Melting Pt: 62…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039212
CAS Number: 886499-21-4