1-Iodo-2-nitro-4-(trifluoromethoxy)benzene
Supplier: Apollo Scientific
Warning
Synonyms: 2-Iodo-5-(trifluoromethoxy)nitrobenzene
PC10109-25G PC10109-5G PC10109-1G
APOSPC10109-25GEA 325 EUR
APOSPC10109-25G APOSPC10109-5G APOSPC10109-1G
1-Iodo-2-nitro-4-(trifluoromethoxy)benzene
1-Iodo-2-nitro-4-(trifluoromethoxy)benzene
Formula: C₇H₃F₃INO₃
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD04039213
CAS Number: 886762-35-2