Ethyl 2,4-bis(trifluoromethyl)imidazo-[1,2-a][1,8]naphthyridine-8-carboxylate
Supplier: Apollo Scientific
PC10117-1G PC10117-5G
APOSPC10117-1GEA 292 EUR
APOSPC10117-1G APOSPC10117-5G
Ethyl 2,4-bis(trifluoromethyl)imidazo-[1,2-a][1,8]naphthyridine-8-carboxylate
Ethyl-2,4-bis(trifluoromethyl)imidazo-[1,2-a][1,8]naphthyridine-8-carboxylate
Formula: C₁₅H₁₁F₆N₃O₂
MW: 379.26 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Ambient
MDL Number: MFCD28385260
CAS Number: 439094-91-4