2-(4-Fluorophenyl)-1-hydrazinecarbothioamide
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Fluorophenyl)thiosemicarbazide
PC10119-1G PC10119-5G
APOSPC10119-1GEA 224 EUR
APOSPC10119-1G APOSPC10119-5G
2-(4-Fluorophenyl)-1-hydrazinecarbothioamide
2-(4-Fluorophenyl)-1-hydrazinecarbothioamide
Formula: C₇H₈FN₃S
MW: 185.22 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Ambient
MDL Number: MFCD01566642
CAS Number: 280133-47-3