3-Methyl-4-[3-(trifluoromethyl)phenyl]-5-isoxazolamine 95%
Supplier: Apollo Scientific
Warning
PC10124-1G PC10124-5G
APOSPC10124-1GEA 259 EUR
APOSPC10124-1G APOSPC10124-5G
3-Methyl-4-[3-(trifluoromethyl)phenyl]-5-isoxazolamine 95%
3-Methyl-4-[3-(trifluoromethyl)phenyl]-5-isoxazolamine
Formula: C₁₁H₉F₃N₂O
MW: 242.2 g/mol
Melting Pt: 120…123 °C
Storage Temperature: Ambient
MDL Number: MFCD03791146
CAS Number: 446276-00-2