3-Nitro-4-{[3-(trifluoromethyl)phenyl]sulphanyl}benzenecarbaldehyde
Supplier: Apollo Scientific
PC10129-1G PC10129-500MG PC10129-5G
APOSPC10129-1GEA 217 EUR
APOSPC10129-1G APOSPC10129-500MG APOSPC10129-5G
3-Nitro-4-{[3-(trifluoromethyl)phenyl]sulphanyl}benzenecarbaldehyde
3-Nitro-4-{[3-(trifluoromethyl)phenyl]sulphanyl}benzenecarbaldehyde
Formula: C₁₄H₈F₃NO₃S
MW: 327.28 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD04117782
CAS Number: 851975-11-6