Ethyl-7-hydroxy-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate
Supplier: Apollo Scientific
PC10147-1G PC10147-500MG PC10147-5G
APOSPC10147-1GEA 275 EUR
APOSPC10147-1G APOSPC10147-500MG APOSPC10147-5G
Ethyl-7-hydroxy-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl-7-hydroxy-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate
Formula: C₁₇H₁₄F₃N₃O₃
MW: 365.31 g/mol
Melting Pt: 273…275 °C
Storage Temperature: Ambient
MDL Number: MFCD03791151
CAS Number: 852690-95-0