2-Chloro-5-(1,3-dihydro-2H-isoindol-2-yl)-4-fluorobenzenol
Supplier: Apollo Scientific
PC10148-1G PC10148-500MG PC10148-5G
APOSPC10148-1GEA 533 EUR
APOSPC10148-1G APOSPC10148-500MG APOSPC10148-5G
2-Chloro-5-(1,3-dihydro-2H-isoindol-2-yl)-4-fluorobenzenol
2-Chloro-5-(1,3-dihydro-2H-isoindol-2-yl)-4-fluorobenzenol
Formula: C₁₄H₁₁ClFNO
MW: 263.7 g/mol
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD03791152
CAS Number: 852690-96-1