4-(Trifluoromethyl)nicotinamide
Supplier: Apollo Scientific
Warning
PC10153-1G PC10153-10G
APOSPC10153-1GEA 36.4 EUR
APOSPC10153-1G APOSPC10153-10G
4-(Trifluoromethyl)nicotinamide
4-(Trifluoromethyl)nicotinamide
Formula: C₇H₅F₃N₂O
MW: 190.1226 g/mol
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00126658
CAS Number: 158062-71-6