2-Oxo-6-phenyl-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
Supplier: Apollo Scientific
Synonyms: 1,2-Dihydro-2-oxo-6-phenyl-4-(trifluoromethyl)pyridine-3-carbonitrile
PC10183-1G PC10183-25G PC10183-5G
APOSPC10183-1GEA 132 EUR
APOSPC10183-1G APOSPC10183-25G APOSPC10183-5G
2-Oxo-6-phenyl-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
2-Oxo-6-phenyl-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
Formula: C₁₃H₇F₃N₂O
MW: 264.21 g/mol
Melting Pt: 300…302 °C
Storage Temperature: Ambient
MDL Number: MFCD00474273
CAS Number: 3335-44-2