4-(4-Chloro-3-fluorophenoxy)benzenecarbonitrile
Supplier: Apollo Scientific
Synonyms: 4-(4-Chloro-3-fluorophenoxy)benzonitrile
PC10188-1G PC10188-500MG PC10188-5G
APOSPC10188-1GEA 172 EUR
APOSPC10188-1G APOSPC10188-500MG APOSPC10188-5G
4-(4-Chloro-3-fluorophenoxy)benzenecarbonitrile
4-(4-Chloro-3-fluorophenoxy)benzenecarbonitrile
Formula: C₁₃H₇ClFNO
MW: 247.66 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD01815095
CAS Number: 449778-68-1