4-(Difluoromethyl)benzoic acid 95%
Supplier: Apollo Scientific
Warning
PC1019-1G PC1019-5G PC1019-250MG
APOSPC1019-1GEA 290 EUR
APOSPC1019-1G APOSPC1019-5G APOSPC1019-250MG
4-(Difluoromethyl)benzoic acid 95%
4-(Difluoromethyl)benzoic acid
Formula: C₈H₆F₂O₂
MW: 172.1289 g/mol
Melting Pt: 203…205 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07784270
CAS Number: 55805-21-5