2-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid
Supplier: Apollo Scientific
Warning
PC10198-1G PC10198-500MG
APOSPC10198-1GEA 85.8 EUR
APOSPC10198-1G APOSPC10198-500MG
2-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid
2-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid
Formula: C₉H₆F₃N₃O₂S
MW: 277.23 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD03012088
CAS Number: 449778-74-9