1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanol
Supplier: Apollo Scientific
PC10201-1G PC10201-500MG PC10201-5G
APOSPC10201-1GEA 405 EUR
APOSPC10201-1G APOSPC10201-500MG APOSPC10201-5G
1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanol
1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanol
Formula: C₇H₈ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD04117790
CAS Number: 959578-12-2