2-(4-Fluorophenoxy)benzenecarbaldehyde oxime
Supplier: Apollo Scientific
PC10206-1G PC10206-500MG
APOSPC10206-1GEA 799 EUR
APOSPC10206-1G APOSPC10206-500MG
2-(4-Fluorophenoxy)benzenecarbaldehyde oxime
2-(4-Fluorophenoxy)benzenecarbaldehyde oxime
Formula: C₁₃H₁₀FNO₂
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD02091015
CAS Number: 449778-79-4