7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carbonitrile
Supplier: Apollo Scientific
PC10212-1G PC10212-500MG
APOSPC10212-1GEA 666 EUR
APOSPC10212-1G APOSPC10212-500MG
7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carbonitrile
7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carbonitrile
Formula: C₁₅H₁₀F₃N₅
Melting Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD03791200
CAS Number: 863305-83-3