1-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-4-ol
Supplier: Apollo Scientific
Warning
PC10225-1G PC10225-500MG PC10225-5G
APOSPC10225-1GEA 314 EUR
APOSPC10225-1G APOSPC10225-500MG APOSPC10225-5G
1-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-4-ol
1-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-4-ol
Formula: C₁₀H₇F₃N₂O
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD03791208
CAS Number: 77458-38-9