3-Chloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline
Supplier: Apollo Scientific
PC10230-1G PC10230-500MG PC10230-5G
APOSPC10230-1GEA 172 EUR
APOSPC10230-1G APOSPC10230-500MG APOSPC10230-5G
3-Chloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline
3-Chloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline
Formula: C₁₂H₇Cl₂F₃N₂O
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD04117811
CAS Number: 76471-06-2