4-(3-Chloro-5-trifluoromethyl-2-pyridyloxy)benzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
PC10233-250MG PC10233-1G PC10233-5G
APOSPC10233-250MGEA 29 EUR
APOSPC10233-250MG APOSPC10233-1G APOSPC10233-5G
4-(3-Chloro-5-trifluoromethyl-2-pyridyloxy)benzenesulphonyl chloride
4-(3-Chloro-5-trifluoromethyl-2-pyridyloxy)benzenesulphonyl chloride
Formula: C₁₂H₆Cl₂F₃NO₃S
MW: 372.139 g/mol
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00243963
CAS Number: 338422-71-2
UN: 3261
ADR: 8,III