4-[2-(4-Bromo-2-fluorophenoxy)phenyl]-3-buten-2-one
Supplier: Apollo Scientific
PC10238-1G PC10238-500MG
APOSPC10238-1GEA 666 EUR
APOSPC10238-1G APOSPC10238-500MG
4-[2-(4-Bromo-2-fluorophenoxy)phenyl]-3-buten-2-one
4-[2-(4-Bromo-2-fluorophenoxy)phenyl]-3-buten-2-one
Formula: C₁₆H₁₂BrFO₂
MW: 335.18 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD02091019
CAS Number: 451485-72-6